missing translation for 'onlineSavingsMsg'
Learn More

(S)-2,2'-Dimethoxy-1,1'-binaphthyl-3,3'-diboronic acid, 97%, Thermo Scientific™

Product Code. 15470416
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15470416 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15470416 Supplier Thermo Scientific Alfa Aesar Supplier No. H53140.MD
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 220204-00-2
Molecular Formula C22H20B2O6
MDL Number MFCD09027249
Quantity 250 mg
Synonym s-2,2'-dimethoxy-1,1'-binaphthalene-3,3'-diboronic acid, r-2,2'-dimethoxy-1,1'-binaphthalene-3,3'-diyl diboronic acid, 2,2'-dimethoxy-1,1'-binaphthalene-3,3'-diyl diboronic acid, 3'-dihydroxyboranyl-2,2'-dimethoxy-1,1'-binaphthalen-3-ylboronic acid, 1,1'-bi 2-methoxy-3-dihydroxyboryl naphthalene, 2,2'-dimethoxy-1,1'-binaphthyl-3,3'-diboronic acid, r-2,2'-dimethoxy-1,1'-binapthalene-3,3'-diboronic acid, s-2,2'-dimethoxy-1,1'-binapthalene-3,3'-diboronic acid, s-2,2'-dimethoxy-1,1'-binaphthyl-3,3'-diyldiboronic acid
InChI Key PNKNNZOHEMBDRL-UHFFFAOYSA-N
SMILES COC1=C(C=C2C=CC=CC2=C1C1=C2C=CC=CC2=CC(B(O)O)=C1OC)B(O)O
IUPAC Name [4-(3-borono-2-methoxynaphthalen-1-yl)-3-methoxynaphthalen-2-yl]boronic acid
Molecular Weight (g/mol) 402.02
PubChem CID 10716074
Formula Weight 402.02
Percent Purity 97%
Chemical Name or Material (S)-2,2'-Dimethoxy-1,1'-binaphthyl-3,3'-diboronic acid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.