missing translation for 'onlineSavingsMsg'
Learn More

(S)-2,2'-Bis(di-p-tolylphosphino)-4,4',6,6'-tetramethoxybiphenyl, 97+%, Thermo Scientific Chemicals

Product Code. 15436106
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1365531-82-3
Molecular Formula C44H44O4P2
Molecular Weight (g/mol) 698.78
MDL Number MFCD19443623
InChI Key JXFDRZQHVPTIMH-UHFFFAOYSA-N
Synonym s-2,2'-bis di-p-tolylphosphino-4,4',6,6'-tetramethoxybiphenyl, r-4,4',6,6'-tetramethoxybiphenyl-2,2'-diyl bis di-p-tolylphosphine, s-4,4',6,6'-tetramethoxybiphenyl-2,2'-diyl bis di-p-tolylphosphine
PubChem CID 87110704

Specifications

Quantity 250 mg
Sensitivity Air sensitive
Formula Weight 698.77
Percent Purity ≥97%
Chemical Name or Material (S)-2,2'-Bis(di-p-tolylphosphino)-4,4',6,6'-tetramethoxybiphenyl

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'