missing translation for 'onlineSavingsMsg'
Learn More

(S)-(-)-2,2'-Bis(di-p-tolylphosphino)-1,1'-binaphthyl, 98%

Product Code. 11329787
Change view
Click to view available options
Quantity:
100 mg
0.5 g
2 g
10 g
Unit Size:
0.5g
100mg
10g
2g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11329787 100 mg 100mg
11339787 0.5 g 0.5g
11306059 2 g 2g
11316059 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11329787 Supplier Thermo Scientific Alfa Aesar Supplier No. 042120.MC
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chiral catalyst ligand | CAS: 100165-88-6 | C48H40P2 | 678.796 g/mol

(S)-(-)-2,2'-Bis(di-p-tolylphosphino)-1,1'-binaphthyl is chiral ligand widely used in asymmetric synthesis. It also used as excellent catalysts for asymmetric hydrogenation of alkenes and some cyclic anhydrides. BINAP is used in organic synthesis for enantioselective transformations catalyzed by its complexes of ruthenium, rhodium, and palladium

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100165-88-6
Molecular Formula C48H40P2
Molecular Weight (g/mol) 678.796
MDL Number MFCD00269856
InChI Key IOPQYDKQISFMJI-UHFFFAOYSA-N
Synonym r-tol-binap, s-tol-binap, s---2,2'-bis di-p-tolylphosphino-1,1'-binaphthyl, 2,2'-bis di-p-tolylphosphino-1,1'-binaphthyl, s-t-binap, rac-tol-binap, r-+-2,2'-bis di-p-tolylphosphino-1,1'-binaphthyl, tol-binap, s-2,2'-bis di-p-tolylphosphino-1,1'-binaphthyl, s-p-tol-binap
PubChem CID 4189906
IUPAC Name [1-[2-bis(4-methylphenyl)phosphanylnaphthalen-1-yl]naphthalen-2-yl]-bis(4-methylphenyl)phosphane
SMILES CC1=CC=C(C=C1)P(C2=CC=C(C=C2)C)C3=C(C4=CC=CC=C4C=C3)C5=C(C=CC6=CC=CC=C65)P(C7=CC=C(C=C7)C)C8=CC=C(C=C8)C

Specifications

Melting Point 255°C to 257°C
Quantity 100 mg
Solubility Information Insoluble in water.
Formula Weight 678.79
Percent Purity 98%
Physical Form Powder
Chemical Name or Material (S)-(-)-2,2'-Bis(di-p-tolylphosphino)-1,1'-binaphthyl

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.