missing translation for 'onlineSavingsMsg'
Learn More

(S)-2-(1-BOC-2-pyrrolidinyl)acetic acid, 97%, Thermo Scientific™

Product Code. 11986631 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11986631 5 g 5g
10275194 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11986631 Supplier Thermo Scientific Acros Supplier No. 399970050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 56502-01-3
Molecular Formula C11H19NO4
Molecular Weight (g/mol) 229.27
MDL Number MFCD01862939
InChI Key GDWKIRLZWQQMIE-QMMMGPOBSA-N
Synonym boc-l-beta-homoproline, s-2-1-tert-butoxycarbonyl pyrrolidin-2-yl acetic acid, boc-beta-hopro-oh, 2s-1-tert-butoxycarbonyl pyrrolidin-2-yl acetic acid, boc-beta3-homopro-oh, s-2-1-boc-2-pyrrolidinyl acetic acid, s-1-boc-2-pyrrolidineacetic acid, 2-pyrrolidineacetic acid, 1-1,1-dimethylethoxy carbonyl-, 2s, 2-2s-1-tert-butoxycarbonylpyrrolidin-2-yl acetic acid, s-2-carboxymethyl-pyrrolidine-1-carboxylic acid tert-butyl ester
PubChem CID 688605
IUPAC Name 2-[(2S)-1-[(2-methylpropan-2-yl)oxycarbonyl]pyrrolidin-2-yl]acetic acid
SMILES CC(C)(C)OC(=O)N1CCCC1CC(=O)O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 97°C to 101°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Quantity 5 g
Specific Rotation −36.6° to −40.6° (20°C, 589nm) (c=1, 4 in DMF)
Formula Weight 229.27
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material (S)-2-(1-BOC-2-pyrrolidinyl)acetic acid
Show More Show Less
compliance-icons
Product Identifier
  • (S)-2-(1-BOC-2-pyrrolidinyl)acetic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.