missing translation for 'onlineSavingsMsg'
Learn More

(S)-2-(1-BOC-2-pyrrolidinyl)acetic acid, 97%, Thermo Scientific™

Product Code. 10275194 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 10275194

Brand: Thermo Scientific Acros 399970010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 56502-01-3
Molecular Formula C11H19NO4
Molecular Weight (g/mol) 229.27
MDL Number MFCD01862939
InChI Key GDWKIRLZWQQMIE-QMMMGPOBSA-N
Synonym boc-l-beta-homoproline, s-2-1-tert-butoxycarbonyl pyrrolidin-2-yl acetic acid, boc-beta-hopro-oh, 2s-1-tert-butoxycarbonyl pyrrolidin-2-yl acetic acid, boc-beta3-homopro-oh, s-2-1-boc-2-pyrrolidinyl acetic acid, s-1-boc-2-pyrrolidineacetic acid, 2-pyrrolidineacetic acid, 1-1,1-dimethylethoxy carbonyl-, 2s, 2-2s-1-tert-butoxycarbonylpyrrolidin-2-yl acetic acid, s-2-carboxymethyl-pyrrolidine-1-carboxylic acid tert-butyl ester
PubChem CID 688605
IUPAC Name 2-[(2S)-1-[(2-methylpropan-2-yl)oxycarbonyl]pyrrolidin-2-yl]acetic acid
SMILES CC(C)(C)OC(=O)N1CCCC1CC(=O)O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 97°C to 101°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 1 g
Specific Rotation −36.6° to −40.6° (20°C, 589nm) (c=1, 4 in DMF)
Formula Weight 229.27
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material (S)-2-(1-BOC-2-pyrrolidinyl)acetic acid
Show More Show Less
compliance-icons
Product Identifier
  • (S)-2-(1-BOC-2-pyrrolidinyl)acetic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.