missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ S 18886
GREENER_CHOICE

Product Code. 18766628
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
This item is not returnable. View return policy

Product Code. 18766628

Brand: Tocris Bioscience™ 5568/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent thromboxane A2 (TP) antagonist

S 18886 is a potent thromboxane A2 (TP) inhibitor (IC50 = 16.4 nM). Inhibits U 46619 (Cat. No. 1932) induced human platelet aggregation and contraction of isolated rabbit saphenous vein in vitro. Also inhibits U 46619 induced increase in tracheal pressure in guinea pigs. Orally available.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (6R)-6-[[(4-Chlorophenyl)sulfonyl]amino]-5,6,7,8-tetrahydro-2-methyl-1-naphthalenepropanoic acid
CAS 165538-40-9
Synonym terutroban
Quantity 10 mg
Target Prostanoid Receptor Antagonists
Molecular Formula C20H22ClNO4S
Formula Weight 407.91
Purity >98%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.