missing translation for 'onlineSavingsMsg'
Learn More

(S)-10-Hydroxycamptothecin, MedChemExpress

Product Code. 16478976
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
100 mg
Unit Size:
100mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16478976 100 mg 100mg
16464747 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16478976 Supplier Medchem Express Supplier No. HYN0095
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress (S)-10-Hydroxycamptothecin (10-HCPT;10-Hydroxycamptothecin) is a DNA topoisomerase I inhibitor of isolated from the Chinese plant Camptotheca accuminata. (S)-10-Hydroxycamptothecin exhibits a remarkable apoptosis-inducing effect. (S)-10-Hydroxycamptothecin has the potential for hepatoma, gastric carcinoma, colon cancer and leukaemia treatment.

(S)-10-Hydroxycamptothecin (10-HCPT

Specifications

Chemical Name or Material (S)-10-Hydroxycamptothecin
CAS 19685-09-7
Color Yellow
Purity Grade Notes Research
Molecular Formula C20H16N2O5
Quantity 100 mg
Synonym 10-HCPT 10-Hydroxycamptothecin
Solubility Information DMSO : 50 mg/mL (137.23 mM; Need ultrasonic)
SMILES O=C1[C@](O)(CC)C2=C(CO1)C(N3CC4=CC5=CC(O)=CC=C5N=C4C3=C2)=O
Molecular Weight (g/mol) 364.35
Formula Weight 364.35
Percent Purity 98.03%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.