missing translation for 'onlineSavingsMsg'
Learn More

(S)-1-[(R)-2-(Diphenylphosphino)ferrocenyl]ethylamine, 97+%, Thermo Scientific Chemicals

Product Code. 15445796
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15445796 250 mg 250mg
15435796 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 15445796 Supplier Thermo Scientific Alfa Aesar Supplier No. H60488.MD
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

(S)-1-[(R)-2-(Diphenylphosphino)ferrocenyl]ethylamine is used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(S)-1-[(R)-2-(Diphenylphosphino)ferrocenyl]ethylamine is used as pharmaceutical intermediates.

Notes
Store in a cool, dry conditions in a well sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 607389-84-4
Molecular Formula C24H24FeNP
Molecular Weight (g/mol) 413.28
MDL Number MFCD11045439
InChI Key DRBVPPSGNFMTKZ-JEZYAYIFNA-N
Synonym (Rp)-1-[(1S)-(1-Aminoethyl)]-2-(diphenylphosphino)ferrocene
PubChem CID 131675344
IUPAC Name cyclopenta-1,3-diene;(1S)-1-(2-diphenylphosphanylcyclopenta-2,4-dien-1-yl)ethanamine;iron
SMILES [Fe].c1cccc1.C[C@H](N)c1cccc1P(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 131°C to 135°C
Quantity 250 mg
Formula Weight 413.27
Percent Purity ≥97%
Chemical Name or Material (S)-1-[(R)-2-(Diphenylphosphino)ferrocenyl]ethylamine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.