missing translation for 'onlineSavingsMsg'
Learn More

(S)-(-)-1-Phenylpropyl isocyanate, 95%

Product Code. 15489149
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15489149 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15489149 Supplier Thermo Scientific Alfa Aesar Supplier No. L20246.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 164033-12-9 | C10H11NO | 161.204 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 164033-12-9
Boiling Point 105°C (17 mmHg)
Molecular Formula C10H11NO
Refractive Index 1.509
MDL Number MFCD05664090
Quantity 1 g
UN Number UN2206
Beilstein 6799586
Sensitivity Moisture sensitive
Synonym s---1-phenylpropyl isocyanate, s-1-phenylpropyl isocyanate, 1s-1-isocyanatopropyl benzene, benzene, 1-isocyanatopropyl-, s, 5-1-isocyanatopropyl benzene, s-1-isocyanatopropyl benzene, benzene, 1s-1-isocyanatopropyl
InChI Key XGPXXSJFZSZULR-JTQLQIEISA-N
Optical Rotation −38.5° (c=1, Chloroform)
SMILES CCC(C1=CC=CC=C1)N=C=O
IUPAC Name [(1S)-1-isocyanatopropyl]benzene
Molecular Weight (g/mol) 161.204
PubChem CID 7018275
Formula Weight 161.2
Percent Purity 95%
Chemical Name or Material (S)-(-)-1-Phenylpropyl isocyanate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.