missing translation for 'onlineSavingsMsg'
Learn More

(S)-(-)-1-Phenylethyl isocyanate, 98%

Product Code. 11317586
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11317586 1 g 1g
11327586 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11317586 Supplier Thermo Scientific Alfa Aesar Supplier No. L11141.03
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 14649-03-7 | C9H9NO | 147.177 g/mol

(S)-(-)-1-Phenylethyl isocyanate act by inhibition of cell proliferation and induction of apoptosis. It helps to prevent lung cancer and esophageal cancer. It is a fine chemical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(S)-(-)-1-Phenylethyl isocyanate act by inhibition of cell proliferation and induction of apoptosis. It helps to prevent lung cancer and esophageal cancer. It is a fine chemical intermediate.

Solubility
Hydrolyzes in water.

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers. Containers which are opened must be carefully resealed and kept upright to prevent leakage. It is sensitive to moisture. Recommended storage temperature is 2 - 8°C.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14649-03-7
Molecular Formula C9H9NO
Molecular Weight (g/mol) 147.177
MDL Number MFCD00002036
InChI Key JJSCUXAFAJEQGB-QMMMGPOBSA-N
Synonym s---1-phenylethyl isocyanate, 1s-1-isocyanatoethyl benzene, s---alpha-methylbenzyl isocyanate, benzene, 1s-1-isocyanatoethyl, isocyanic acid s---a-methylbenzyl ester, s-alpha-methylbenzyl isocyanate, s---1-phenylethylisocyanate, isocyanic acid s---alpha-methylbenzyl ester, isocyanic acid s---methylbenzyl ester, 1-isocyanatoethyl benzene #
PubChem CID 2724173
IUPAC Name [(1S)-1-isocyanatoethyl]benzene
SMILES CC(C1=CC=CC=C1)N=C=O

Specifications

Density 1.045
Boiling Point 55°C to 56°C (2.5 mmHg)
Flash Point 65°C (149°F)
Odor Pungent
Refractive Index 1.5145
Quantity 1 g
UN Number UN2206
Beilstein 4230972
Sensitivity Moisture sensitive; May form a precipitate
Solubility Information Hydrolyzes in water.
Optical Rotation −10.5° (Neat)
Formula Weight 147.18
Percent Purity 98%
Chemical Name or Material (S)-(-)-1-Phenylethyl isocyanate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.