missing translation for 'onlineSavingsMsg'
Learn More

(S)-(-)-1-Phenyl-1-propanol, TRC

Product Code. 30413265
Change view
Click to view available options
Quantity:
50 mg
100 mg
250 mg
Unit Size:
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30413265 100 mg -
30417798 250 mg -
30416525 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30413265 Supplier Toronto Research Chemicals Supplier No. TRCP336188100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 613-87-6
Molecular Formula C9H12O
Molecular Weight (g/mol) 136.19
Synonym (-)-1-Phenylpropan-1-ol, (-)-1-Phenylpropanol, (-)-1-Phenylpropyl Alcohol, (-)-alpha-Ethylbenzyl Alcohol, (-)-alpha-Phenylpropanol, (1S)-1-Phenyl-1-propanol, (S)-(-)-1-Phenyl-1-propanol, (S)-1-Phenylpropan-1-ol, (S)-1-Phenylpropanol, (S)-Phenylethylcarbinol, (S)-alpha-Ethylbenzenemethanol, (S)-alpha-Ethylbenzyl alcohol, (S)-alpha-Hydroxypropylbenzene
IUPAC Name (1S)-1-phenylpropan-1-ol
SMILES CC[C@H](O)c1ccccc1

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C9H12O/c1-2-9(10)8-6-4-3-5-7-8/h3-7,9-10H,2H2,1H3/t9-/m0/s1
Formula Weight 136.0888
Chemical Name or Material (S)-(-)-1-Phenyl-1-propanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.