missing translation for 'onlineSavingsMsg'
Learn More

(S)-(-)-1-Octyn-3-ol, 98+%, Thermo Scientific Chemicals

Product Code. p-7047373
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11379585 250 mg 250mg
11369585 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11379585 Supplier Thermo Scientific Alfa Aesar Supplier No. L19046.MD

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 32556-71-1 | Molecular Formula: C8H14O | Molecular Weight: 126.199 g/mol

It is a key intermediate for the synthesis of prostaglandins. It is used in the synthesis of (R)-3-aminooctanoic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is a key intermediate for the synthesis of prostaglandins. It is used in the synthesis of (R)-3-aminooctanoic acid.

Notes
Store away from strong oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Keep ignition sources away.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 32556-71-1
Molecular Formula C8H14O
Molecular Weight (g/mol) 126.199
MDL Number MFCD00191475
InChI Key VUGRNZHKYVHZSN-MRVPVSSYSA-N
Synonym s-1-octyn-3-ol, s---1-octyn-3-ol, s-3-hydroxy-1-octyne, 1-octyn-3-ol, 3s, 3s-1-octyn-3-ol, unii-d8ho63xx92, 3s-oct-1-yn-3-ol, 3s-hydroxy-1-octyne, --1-octyn-3-ol, 1-octyn-3-ol, s
PubChem CID 2734534
IUPAC Name (3S)-oct-1-yn-3-ol
SMILES CCCCCC(C#C)O

Specifications

Density 0.866
Boiling Point 100°C (20 mmHg)
Flash Point 63°C (145°F)
Refractive Index 1.442
Quantity 250 mg
Beilstein 2037703
Optical Rotation −9.9° (Neat)
Formula Weight 126.2
Percent Purity ≥98%
Chemical Name or Material (S)-(-)-1-Octyn-3-ol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.