missing translation for 'onlineSavingsMsg'
Learn More

(S)-(+)-1-Methoxy-2-propanol, 99%

Product Code. 10370114 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 mL
5 mL
25 mL
Packaging:
Glass bottle
Unit Size:
1mL
25mL
5mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10370114 25 mL Glass bottle 25mL
10011851 1 mL Glass bottle 1mL
10726432 5 mL Glass bottle 5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10370114 Supplier Thermo Scientific Acros Supplier No. 331790250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 26550-55-0 | C4H10O2 | 90.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 26550-55-0
Molecular Formula C4H10O2
Molecular Weight (g/mol) 90.12
MDL Number MFCD01632588
InChI Key ARXJGSRGQADJSQ-BYPYZUCNSA-N
Synonym s-+-1-methoxy-2-propanol, 2s-1-methoxypropan-2-ol, s-1-methoxypropan-2-ol, s---1-methoxy-2-propanol, s-1-methoxy-2-propanol, 2-propanol, 1-methoxy-, 2s, 2s-1-methoxy-2-propanol, pubchem6715, s-1-methoxy-propan-2-ol
PubChem CID 6993459
IUPAC Name (2S)-1-methoxypropan-2-ol
SMILES CC(COC)O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point -97.0°C
Color Colorless
Density 0.9200g/mL
Boiling Point 120.0°C
Flash Point 30°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3OCH2CH(OH)CH3
Refractive Index 1.4020 to 1.4040
Quantity 25 mL
Specific Gravity 0.92
Specific Rotation Condition +20° (20°C c=10,CHCl3)
Specific Rotation + 20.00
Solubility Information Solubility in water: soluble
Formula Weight 90.12
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material (S)-(+)-1-Methoxy-2-propanol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.