missing translation for 'onlineSavingsMsg'
Learn More

(S)-1-Cyclopropylethylamine, ChiPros™, 98%, ee 98+%

Product Code. 11491740
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11491740 1 g 1g
11401750 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11491740 Supplier Thermo Scientific Alfa Aesar Supplier No. H27499.03
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 195604-39-8 | C5H11N | 85.15 g/mol

(S)-1-Cyclopropylethylamine acts as a chiral and organic building block in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(S)-1-Cyclopropylethylamine acts as a chiral and organic building block in organic synthesis.

Solubility
Miscible with water.

Notes
Air sensitive. Incompatible with acids and strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 195604-39-8
Molecular Formula C5H11N
Molecular Weight (g/mol) 85.15
MDL Number MFCD08064289
InChI Key IXCXVGWKYIDNOS-BYPYZUCNSA-N
PubChem CID 7363995
IUPAC Name (1S)-1-cyclopropylethanamine
SMILES CC(C1CC1)N

Specifications

Melting Point -68°C
Density 0.799
Boiling Point 95°C
Flash Point 3°C (37°F)
Quantity 1 g
UN Number UN2733
Sensitivity Air Sensitive
Synonym s-1-cyclopropylethylamine, s-1-cyclopropylethanamine, 1s-1-cyclopropylethanamine, 1s-1-cyclopropylethan-1-amine, 1 s-1-cyclopropylethanamine, n-s-1-cyclopropyl-ethyl-amine, s-1-cyclopropylethylamine, chipros, cyclopropanemethanamine,a-methyl-, as, s-1-cyclopropylethylamine, chipros, ee 98+%, s-1-cyclopropylethylamine, chipros r , produced by basf
Solubility Information Miscible with water.
Formula Weight 85.15
Percent Purity 98%
Assay ee ≥98%
Chemical Name or Material (S)-1-Cyclopropylethylamine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.