missing translation for 'onlineSavingsMsg'
Learn More

(S)-(+)-1-Cyclohexylethyl isocyanate, 94%, Thermo Scientific™

Product Code. 15459139
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15459139 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15459139 Supplier Thermo Scientific Alfa Aesar Supplier No. L20229.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 93470-27-0
Boiling Point 210°C to 215°C
Molecular Formula C9H15NO
Refractive Index 1.465
MDL Number MFCD05664074
Quantity 1 g
UN Number UN2206
Beilstein 9255849
Sensitivity Moisture sensitive
Synonym s-+-1-cyclohexylethyl isocyanate, 1s-1-isocyanatoethyl cyclohexane, 1s-1-cyclohexylethanisocyanate, s-1-cyclohexylethyl isocyanate, s-+-1-cyclohexylethylisocyanate
InChI Key IGSKYRAPJLTXSO-QMMMGPOBSA-N
SMILES CC(C1CCCCC1)N=C=O
IUPAC Name [(1S)-1-isocyanatoethyl]cyclohexane
Molecular Weight (g/mol) 153.225
PubChem CID 40427464
Formula Weight 153.23
Percent Purity 94%
Chemical Name or Material (S)-(+)-1-Cyclohexylethyl isocyanate
Show More Show Less
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.