missing translation for 'onlineSavingsMsg'
Learn More

(S)-(-)-1-Boc-4-oxopiperidine-2-carboxylic acid, 95%, Thermo Scientific Chemicals

Product Code. 15443556
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15443556 1 g 1g
15453556 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15443556 Supplier Thermo Scientific Alfa Aesar Supplier No. H57617.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

It is a fine chemical and synthetic intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is a fine chemical and synthetic intermediate.

Solubility
Soluble (12 g/L) (25°C)

Notes
Store in a cool ventilated place. Avoid strong oxidizing materials. Hygroscopic.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 198646-60-5
Molecular Formula C11H17NO5
Molecular Weight (g/mol) 243.259
MDL Number MFCD03094867
InChI Key GPBCBXYUAJQMQM-QMMMGPOBSA-N
Synonym s-1-tert-butoxycarbonyl-4-oxopiperidine-2-carboxylic acid, s-1-boc-4-oxopipecolic acid, s---1-boc-4-oxopiperidine-2-carboxylic acid, s-1-boc-4-oxopiperidine-2-carboxylic acid, 2s-1-tert-butoxycarbonyl-4-oxopiperidine-2-carboxylic acid, 2s-1-tert-butoxy carbonyl-4-oxopiperidine-2-carboxylic acid, s-1-tert-butoxycarbonyl-4-oxopiperidine-2-carboxylicacid, 2s-4-oxo-1,2-piperidinedicarboxylic acid 1-1,1-dimethylethyl ester, 2s-n-boc-4-oxopipecolic acid, r-+-1-boc-4-oxopiperidine-2-carboxylic acid
PubChem CID 10857681
IUPAC Name (2S)-1-[(2-methylpropan-2-yl)oxycarbonyl]-4-oxopiperidine-2-carboxylic acid
SMILES CC(C)(C)OC(=O)N1CCC(=O)CC1C(=O)O

Specifications

Melting Point 121°C to 126°C
Quantity 1 g
Sensitivity Air and moisture sensitive
Solubility Information Soluble (12g/L) (25°C)
Optical Rotation −22° (c=1 in Chloroform)
Formula Weight 243.26
Percent Purity 95%
Chemical Name or Material (S)-(-)-1-Boc-4-oxopiperidine-2-carboxylic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.