missing translation for 'onlineSavingsMsg'
Learn More

(S)-1-BOC-3-methylpiperazine, 98%, ACROS Organics™

Product Code. 10165947 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10165947 1 g Glass bottle 1g
10789228 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10165947 Supplier Thermo Scientific Acros Supplier No. 429700010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 147081-29-6
Molecular Formula C10H20N2O2
Molecular Weight (g/mol) 200.282
MDL Number MFCD02683204
InChI Key FMLPQHJYUZTHQS-QMMMGPOBSA-N
Synonym s-4-n-boc-2-methylpiperazine, s-4-n-boc-2-methyl piperazine, s-tert-butyl 3-methylpiperazine-1-carboxylate, s-4-boc-2-methylpiperazine, s-1-boc-3-methyl-piperazine, tert-butyl 3s-3-methylpiperazine-1-carboxylate, s-1-boc-3-methylpiperazine, s-4bocmp, 4-boc-2-s-methyl-piperazine, s-1-n-boc-3-methylpiperazine
PubChem CID 7023035
IUPAC Name tert-butyl (3S)-3-methylpiperazine-1-carboxylate
SMILES CC1CN(CCN1)C(=O)OC(C)(C)C

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 45.0°C to 48.5°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 1 g
Specific Rotation −23° to −27° (20°C, 589nm) (c=2.5, MeOH)
Formula Weight 200.28
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material (S)-1-BOC-3-Methylpiperazine
Show More Show Less
compliance-icons
Product Identifier
  • (S)-1-Boc-3-methylpiperazine
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.