missing translation for 'onlineSavingsMsg'
Learn More

(S)-1-Boc-3-hydroxypiperidine, 97%

Product Code. 15429507
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 15429507

Brand: Thermo Scientific Alfa Aesar H52771.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 143900-44-1 | C10H19NO3 | 201.27 g/mol

(S)-1-Boc-3-hydroxypiperidine is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(S)-1-Boc-3-hydroxypiperidine is used as pharmaceutical intermediate.

Solubility
Soluble in methanol.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 143900-44-1
Molecular Formula C10H19NO3
Molecular Weight (g/mol) 201.27
MDL Number MFCD04115307
InChI Key UIJXHKXIOCDSEB-UHFFFAOYNA-N
Synonym s-1-boc-3-hydroxypiperidine, s-tert-butyl 3-hydroxypiperidine-1-carboxylate, s-n-boc-3-hydroxypiperidine, tert-butyl 3s-3-hydroxypiperidine-1-carboxylate, s-1-n-boc-3-hydroxy-piperidine, s-1-n-boc-3-hydroxypiperidine, s-n-boc-3-hydroxy piperidine, s-3-hydroxypiperidine, n-boc protected, s-3-hydroxy-piperidine-1-carboxylic acid tert-butyl ester, 3s-1-tert-butoxycarbonyl-3-hydroxypiperidine
PubChem CID 1514399
IUPAC Name tert-butyl 3-hydroxypiperidine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCCC(O)C1

Specifications

Melting Point 46°C to 50°C
Flash Point >110°C (230°F)
Quantity 1 g
Solubility Information Soluble in methanol.
Optical Rotation +23° (c=5 in Methanol)
Formula Weight 201.27
Percent Purity 97%
Chemical Name or Material (S)-1-Boc-3-hydroxypiperidine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.