missing translation for 'onlineSavingsMsg'
Learn More

(S)-(-)-1-BOC-3-aminopyrrolidine, 95%

Product Code. 10637030 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10637030 1 g Glass bottle 1g
10783342 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10637030 Supplier Thermo Scientific Acros Supplier No. 435450010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 147081-44-5 | C9H18N2O2 | 186.26 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 147081-44-5
Molecular Formula C9H18N2O2
Molecular Weight (g/mol) 186.26
MDL Number MFCD03419271
InChI Key CMIBWIAICVBURI-UHFFFAOYNA-N
Synonym s-1-boc-3-aminopyrrolidine, s-3-amino-1-n-boc-pyrrolidine, s---1-boc-3-aminopyrrolidine, tert-butyl 3s-3-aminopyrrolidine-1-carboxylate, s-tert-butyl 3-aminopyrrolidine-1-carboxylate, s-3-amino-n-boc-pyrrolidine, s-bocap, s---1-tert-butoxycarbonyl-3-aminopyrrolidine, s-3-amino-1-boc-pyrrolidine, s---n-boc-3-aminopyrrolidine
PubChem CID 854071
IUPAC Name tert-butyl (3S)-3-aminopyrrolidine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCC(N)C1

Specifications

CAS Min % 94.0
CAS Max % 100.0
Color Colorless to Yellow
Boiling Point 120.0°C to 125.0°C (5.0 Torr)
Flash Point 91°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Quantity 1 g
Specific Rotation -1.5° to -3.0° (20°C, 589nm) (C=1, chloroform)
Formula Weight 186.25
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material (S)-(-)-1-BOC-3-aminopyrrolidine
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • (S)-(-)-1-Boc-3-aminopyrrolidine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H318-Causes serious eye damage.
  • H301-Toxic if swallowed.
  • EUH044-Risk of explosion if heated under confinement.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.