missing translation for 'onlineSavingsMsg'
Learn More

(S)-(-)-1-Boc-3-aminopyrrolidine, 95%

Product Code. 11305535
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11305535 5 g 5g
11395525 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11305535 Supplier Thermo Scientific Alfa Aesar Supplier No. L19696.06
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 147081-44-5 | C9H18N2O2 | 186.26 g/mol

(S)-(-)-1-Boc-3-aminopyrrolidine is specifically used to protect amine in the solid phase synthesis of peptides. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(S)-(-)-1-Boc-3-aminopyrrolidine is specifically used to protect amine in the solid phase synthesis of peptides. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.

Notes
Store under inert gas. It is sensitive to air. Store in a cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 147081-44-5
Molecular Formula C9H18N2O2
Molecular Weight (g/mol) 186.26
MDL Number MFCD03419271
InChI Key CMIBWIAICVBURI-UHFFFAOYNA-N
Synonym s-1-boc-3-aminopyrrolidine, s-3-amino-1-n-boc-pyrrolidine, s---1-boc-3-aminopyrrolidine, tert-butyl 3s-3-aminopyrrolidine-1-carboxylate, s-tert-butyl 3-aminopyrrolidine-1-carboxylate, s-3-amino-n-boc-pyrrolidine, s-bocap, s---1-tert-butoxycarbonyl-3-aminopyrrolidine, s-3-amino-1-boc-pyrrolidine, s---n-boc-3-aminopyrrolidine
PubChem CID 854071
SMILES CC(C)(C)OC(=O)N1CCC(N)C1

Specifications

Density 1.02
Boiling Point 115°C (4 mmHg)
Refractive Index 1.4715
Quantity 5 g
UN Number UN2735
Beilstein 8312125
Sensitivity Air sensitive
Optical Rotation −3° (Neat)
IUPAC Name tert-butyl (3S)-3-aminopyrrolidine-1-carboxylate
Formula Weight 186.25
Percent Purity 98%
Assay ee 99%
Chemical Name or Material (S)-(-)-1-Boc-3-aminopyrrolidine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.