missing translation for 'onlineSavingsMsg'
Learn More

(S)-1-BOC-3-(Aminomethyl)piperidine, 97%, ACROS Organics™

Brand:  Acros Organics 437470050

Code : 29

Additional Details : CAS Number : 140645-24-5 Weight : 0.06000kg


Quantity 5g

 View more versions of this product

Product Code. 10159829

  • £422.20 / 5g


EU Stock 6
Add to basket


Specifications

Chemical Identifiers

(S)-1-BOC-3-(Aminomethyl)piperidine
96.0
≥96.0%
C11H22N2O2
140645-24-5
100.0
97%
214.31

Specifications

(S)-1-BOC-3-(Aminomethyl)piperidine
96.0
Authentic
-10° to -20° (20°C, 589nm) (c=1, methanol)
97%
214.31
Clear colorless
140645-24-5
100.0
Liquid
≥96.0%
C11H22N2O2
5g
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • (S)-1-N-BOC-3-(Aminomethyl)piperidine

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

SDS