missing translation for 'onlineSavingsMsg'
Learn More

(S)-(-)-1-Boc-2-cyanopyrrolidine, 95%, Thermo Scientific™

Product Code. 15493688
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15493688 1 g 1g
15403698 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15493688 Supplier Thermo Scientific Alfa Aesar Supplier No. H57150.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
(S)-(-)-1-Boc-2-pyrrolidinecarbonitrile is used in synthesis of prolyl oligopeptidase inhibitors and is a reagent that is used in the design of potent and selective inhibitors of DPP-4 that exhibits an attractive pharmacokinetic profile as antidiabetics.

Solubility
Slightly soluble in water.

Notes
Moisture and light sensitive. Store away from light, water/moisture and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store in dark.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 228244-04-0
Molecular Formula C10H16N2O2
Molecular Weight (g/mol) 196.25
MDL Number MFCD01861224
InChI Key MDMSZBHMBCNYNO-UHFFFAOYNA-N
Synonym s-1-boc-2-cyanopyrrolidine, s-tert-butyl 2-cyanopyrrolidine-1-carboxylate, s-1-n-boc-2-cyano-pyrrolidine, tert-butyl 2s-2-cyanopyrrolidine-1-carboxylate, s-1-boc-2-cyano-pyrrolidine, s-1-n-boc-2-cyanopyrrolidine, s-1-boc-2-pyrrolidinecarbonitrile, s-n-boc-pyrrolidine-2-carbonitrile, s---1-boc-2-cyanopyrrolidine, s-n-boc-2-pyrrolidinonitrile
PubChem CID 6951263
IUPAC Name tert-butyl (2S)-2-cyanopyrrolidine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCCC1C#N

Specifications

Melting Point 33°C to 36°C
Flash Point 110°C (230°F)
Quantity 1 g
Sensitivity Light and moisture sensitive
Solubility Information Slightly soluble in water.
Optical Rotation +105° (c=1 in Chloroform)
Formula Weight 196.25
Percent Purity 95%
Chemical Name or Material (S)-(-)-1-Boc-2-cyanopyrrolidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.