missing translation for 'onlineSavingsMsg'
Learn More

(S)-1-Beta-Hydroxy-1-(2-thienyl)-3-methylaminopropane, TRC

Product Code. 30136241
Change view
Click to view available options
Quantity:
10 mg
100 mg
1 g
Unit Size:
100mg
10mg
1g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30136241 1 g 1g
30135928 100 mg 100mg
30008258 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30136241 Supplier Toronto Research Chemicals Supplier No. TRCH9618151G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 116539-55-0
Molecular Formula C8 H13 N O S
Molecular Weight (g/mol) 171.26
Synonym 2-Thiophenemethanol, α-[2-(methylamino)ethyl]-, (S)-, (1S)-(-)-3-(Methylamino)-1-(thien-2-yl)propan-1-ol, (S)-(-)-3-(N-Methylamino)-1-(2-thienyl)-1-propanol, (S)-(-)-3-Methylamino-1-(2-thienyl)propan-1-ol, (S)-3-Methylamino-1-(2-thienyl)-1-propanol, N-Methyl-(S)-(-)-3-Hydroxy-3-(2-thiophen)propylamine, (1S)-3-(Methylamino)-1-(thiophen-2-yl)propan-1-ol
IUPAC Name (1S)-3-(methylamino)-1-thiophen-2-ylpropan-1-ol
SMILES CNCC[C@H](O)c1cccs1

Specifications

Quantity 1 g
InChI Formula InChI=1S/C8H13NOS/c1-9-5-4-7(10)8-3-2-6-11-8/h2-3,6-7,9-10H,4-5H2,1H3/t7-/m0/s1
Formula Weight 171.0718
Chemical Name or Material (1S)-3-(Methylamino)-1-(thiophen-2-yl)propan-1-ol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.