missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(S)-1-Benzyl-3-piperidinol, Mikromol
Click to view available options
Quantity:
100 mg
Description
(S)-1-Benzyl-3-piperidinol, Mikromol
Specifications
Specifications
| Chemical Name or Material | (S)-1-Benzyl-3-piperidinol |
| CAS | 91599-79-0 |
| Analyte or Component Names | (S)-1-Benzyl-3-piperidinol |
| Shipping Condition | Room Temperature |
| Molecular Formula | C12 H17 N O |
| Certifications/Compliance | ISO 9001 |
| Synonym | 3-Piperidinol, 1-(phenylmethyl)-, (3S)-, 3-Piperidinol, 1-(phenylmethyl)-, (S)-, (3S)-1-(Phenylmethyl)-3-piperidinol, (S)-1-Benzylpiperidin-3-ol |
| InChI Formula | InChI=1S/C12H17NO/c14-12-7-4-8-13(10-12)9-11-5-2-1-3-6-11/h1-3,5-6,12,14H,4,7-10H2/t12-/m0/s1 |
| Solution Type | Neat |
| SMILES | O[C@H]1CCCN(Cc2ccccc2)C1 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction