missing translation for 'onlineSavingsMsg'
Learn More

(S)-(+)-1-Benzyl-3-aminopyrrolidine, 99%, Thermo Scientific™

Product Code. 10881691 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10881691 1 g Glass bottle 1g
10483520 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10881691 Supplier Thermo Scientific Acros Supplier No. 437880010
 Restricted Item
Estimated Shipment: 07-08-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 114715-38-7
Molecular Formula C11H16N2
Molecular Weight (g/mol) 176.26
InChI Key HBVNLKQGRZPGRP-NSHDSACASA-N
Synonym s-+-1-benzyl-3-aminopyrrolidine, 3s-1-benzylpyrrolidin-3-amine, 3s-+-1-benzyl-3-aminopyrrolidine, s-n-benzyl-3-aminopyrrolidine, s-3-amino-1-n-benzyl-pyrrolidine, s-1-benzylpyrrolidin-3-amine, s-1-benzyl-3-aminopyrrolidine, s-3-amino-1-benzylpyrrolidine, s-bap, s-3-amino-1-n-benzylpyrrolidine
PubChem CID 1519353
IUPAC Name (3S)-1-benzylpyrrolidin-3-amine
SMILES C1CN(CC1N)CC2=CC=CC=C2

Specifications

CAS Min % 98.5
CAS Max % 100.0
Color Colorless to Yellow
Density 1.0200g/mL
Boiling Point 108°C to 115°C (5.0 mmHg)
Flash Point 140°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Quantity 1 g
Specific Gravity 1.02
Specific Rotation + 2
Formula Weight 176.26
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material (S)-(+)-1-Benzyl-3-aminopyrrolidine
Show More Show Less
compliance-icons
Product Identifier
  • (S)-(+)-1-Benzyl-3-aminopyrrolidine
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.