missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ (S)-(+)-1-Aminoindane, ChiPros 99+%, ee 99+%

Product Code. 11394196
Change view
Click to view available options
Quantity:
1g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11394196 1g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11394196 Supplier Alfa Aesar™ Supplier No. L19303.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Applications
(S)-(+)-1-Aminoindane is the S-enantiomer of 1-aminoindane, which is the metabolite of the monoamine oxidase-B inhibitor rasagiline. It is also used as a chiral and organic building block.

Solubility
Miscible with toluene. Immiscible with water.

Notes
Incompatible with strong oxidizing agents and carbondioxide
TRUSTED_SUSTAINABILITY

Specifications

CAS 61341-86-4
Density 1.038
Boiling Point 225°C
Flash Point 94°C (201°F)
Molecular Formula C9H11N
Refractive Index 1.562
MDL Number MFCD00216670
Quantity 1g
Beilstein 2206708
Sensitivity Air Sensitive
Synonym s-+-1-aminoindan, s-1-aminoindane, s-2,3-dihydro-1h-inden-1-amine, s-+-1-aminoindane, 1s-2,3-dihydro-1h-inden-1-amine, s-+-1-indanamine, 1s-indan-1-amine, s-indan-1-ylamine, 1h-inden-1-amine, 2,3-dihydro-, 1s
Solubility Information Miscible with toluene. Immiscible with water.
InChI Key XJEVHMGJSYVQBQ-UHFFFAOYNA-N
Optical Rotation +16.5° (c=1.5 in Methanol)
SMILES NC1CCC2=CC=CC=C12
IUPAC Name (1S)-2,3-dihydro-1H-inden-1-amine
Molecular Weight (g/mol) 133.19
PubChem CID 7000084
Formula Weight 133.19
Percent Purity ≥99%
Assay ee ≥99%
Chemical Name or Material (S)-(+)-1-Aminoindane
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.