missing translation for 'onlineSavingsMsg'
Learn More

(S)-(+)-1-Amino-2-propanol, 98+%

Product Code. 10510014 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10510014 25 g Glass bottle 25g
10022930 1 g Glass bottle 1g
10500014 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10510014 Supplier Thermo Scientific Acros Supplier No. 218980250
 Restricted Item
Estimated Shipment: 20-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 2799-17-9 | C3H9NO | 75.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2799-17-9
Molecular Formula C3H9NO
Molecular Weight (g/mol) 75.11
MDL Number MFCD00064429
InChI Key HXKKHQJGJAFBHI-UHFFFAOYNA-N
Synonym s-+-1-amino-2-propanol, s-1-aminopropan-2-ol, 2s-1-aminopropan-2-ol, +-isopropanolamine, s-1-amino-2-propanol, 2-propanol, 1-amino-, 2s, 1-amino-2-propanol #, 2-propanol, 1-amino-, s, ambotzfal1033
PubChem CID 7311736
IUPAC Name 1-aminopropan-2-ol
SMILES CC(O)CN

Specifications

CAS Min % 98.0
CAS Max % 100.0
Melting Point 19°C to 21°C
Color Colorless to Yellow
Density 0.9540g/mL
Boiling Point 160°C
Flash Point 71°C
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Packaging Glass bottle
Linear Formula CH3CH(OH)CH2NH2
Refractive Index 1.4465 to 1.4495
Quantity 25 g
Beilstein 04, IV, 1665
Specific Gravity 0.954
Specific Rotation Condition + 18.00 (20.00°C c=1.8,H2O)
Specific Rotation + 18
Solubility Information Solubility in water: soluble. Other solubilities: soluble in organic solvents
Formula Weight 75.11
Percent Purity 98+%
Physical Form Liquid After Melting
Chemical Name or Material (S)-(+)-1-Amino-2-propanol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.