missing translation for 'onlineSavingsMsg'
Learn More

(S)-(-)-1-(4-Methoxyphenyl)ethyl isocyanate, 95%

Product Code. 15439149
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15439149 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15439149 Supplier Thermo Scientific Alfa Aesar Supplier No. L20240.03
 Restricted Item
Estimated Shipment: 16-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 745783-83-9 | C10H11NO2 | 177.203 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 745783-83-9
Molecular Formula C10H11NO2
Refractive Index 1.518
MDL Number MFCD05664085
Quantity 1 g
UN Number UN2206
Sensitivity Moisture sensitive
Synonym s---1-4-methoxyphenyl ethyl isocyanate, 1-1s-1-isocyanatoethyl-4-methoxybenzene, s-1-4-methoxyphenyl ethylisocyanate, s---1-4-methoxyphenyl ethylisocyanate, s ?-?-? ?-?1-? 4-?methoxyphenyl ?ethyl isocyanat, benzene, 1-1s-1-isocyanatoethyl-4-methoxy-9ci, s---1-3-methoxyphenyl ethyl isocyanate
InChI Key WTNKKJUXEYSETC-QMMMGPOBSA-N
SMILES CC(C1=CC=C(C=C1)OC)N=C=O
IUPAC Name 1-[(1S)-1-isocyanatoethyl]-4-methoxybenzene
Molecular Weight (g/mol) 177.203
PubChem CID 7018272
Formula Weight 177.2
Percent Purity 95%
Chemical Name or Material (S)-(-)-1-(4-Methoxyphenyl)ethyl isocyanate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.