missing translation for 'onlineSavingsMsg'
Learn More

(S)-(+)-1,4-Dihydro-2,6-dimethyl-4-(3-nitrophenyl)-3,5-pyridinedicarboxylic Acid Monomethyl Ester, TRC

Product Code. 30385471
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30385471 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30385471 Supplier Toronto Research Chemicals Supplier No. TRCD44912110MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifikationer

Chemical Name or Material (S)-(+)-1,4-Dihydro-2,6-dimethyl-4-(3-nitrophenyl)-3,5-pyrid
CAS 76093-34-0
Molecular Formula C16 H16 N2 O6
Quantity 10 mg
InChI Formula InChI=1S/C16H16N2O6/c1-8-12(15(19)20)14(13(9(2)17-8)16(21)24-3)10-5-4-6-11(7-10)18(22)23/h4-7,14,17H,1-3H3,(H,19,20)/t14-/m0/s1
SMILES COC(=O)C1=C(C)NC(=C([C@@H]1c2cccc(c2)[N+](=O)[O-])C(=O)O)C
IUPAC Name (4S)-5-methoxycarbonyl-2,6-dimethyl-4-(3-nitrophenyl)-1,4-dihydropyridine-3-carboxylic acid
Molecular Weight (g/mol) 332.31
Formula Weight 332.1008
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.