missing translation for 'onlineSavingsMsg'
Learn More

(S)-1,4-Diazabicyclo[4.3.0]nonane, 98%, Thermo Scientific™

Product Code. 15452182 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15452182 250 mg 250mg
15442182 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15452182 Supplier Thermo Scientific Acros Supplier No. 456812500

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 93643-24-4
Molecular Formula C7H14N2
Molecular Weight (g/mol) 126.20
MDL Number MFCD03787926
InChI Key FTTATHOUSOIFOQ-SSDOTTSWSA-N
Synonym s-octahydropyrrolo 1,2-a pyrazine, s-1,4-diazabicyclo 4.3.0 nonane, 8as-octahydropyrrolo 1,2-a pyrazine, 8as-octahydropyrrolo 1,2-a piperazine, s-octahydro-pyrrolo 1,2-a pyrazine, 6s-1,4-diazabicyclo 4.3.0 nonane, pyrrolo 1,2-a pyrazine, octahydro-, 8as, 8as-1,2,3,4,6,7,8,8a-octahydropyrrolo 1,2-a pyrazine, pubchem20597, 6s-1,4-diazabicyclo 4,3,0 nonane
PubChem CID 781249
IUPAC Name (8aS)-1,2,3,4,6,7,8,8a-octahydropyrrolo[1,2-a]pyrazine
SMILES C1C[C@@H]2CNCCN2C1

Specifications

Flash Point 89°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Quantity 250 mg
Specific Rotation Condition + 5.88 (20.00°C c=1, CHCl3)
Specific Rotation + 5.88
Formula Weight 126.2
Percent Purity 98%
Chemical Name or Material (S)-1,4-Diazabicyclo[4.3.0]nonane
compliance-icons
Product Identifier
  • (S)-1,4-Diazabicyclo[4.3.0]nonane
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.