missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(S)-1-(4-Chlorophenyl)ethylamine, 95+%, (sum of isomers), 95% e.e., Thermo Scientific™
Shop All Thermo Scientific Chemicals ProductsDescription
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Specifications
| Density | 1.1150g/mL |
| Quantity | 5 g |
| Packaging | Glass bottle |
| Refractive Index | 1.5410 to 1.5430 |
| Assay Percent Range | 95% min. (GC) |
| Specific Gravity | 1.115 |
| Linear Formula | ClC6H4CH(CH3)NH2 |
| Boiling Point | 232.0°C |
| Solubility Information | Solubility in water: moderately soluble |
| Formula Weight | 155.63 |
| Show More |
Safety and Handling
Product Identifier
- (S)-1-(4-Chlorophenyl)ethylamine
- Danger
- Acute toxicity Category 2
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 2
- Serious eye damage/eye irritation Category 1
- Respiratory sensitiser Category 1
- Skin corrosion/irritation Category 2
- H317-May cause an allergic skin reaction.
- H318-Causes serious eye damage.
- H315-Causes skin irritation.
- H330-Fatal if inhaled.
- H411-Toxic to aquatic life with long lasting effects.
- H302-Harmful if swallowed.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?