missing translation for 'onlineSavingsMsg'
Learn More

(S)-(-)-1-(4-Bromophenyl)ethylamine, ChiPros, 99%, ee 98%

Product Code. 11314516
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11314516 5 g 5g
11304516 1 g 1g
11324516 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11314516 Supplier Thermo Scientific Alfa Aesar Supplier No. L20080.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 27298-97-1 | C8H11BrN | 201.09 g/mol

(S)-(-)-1-(4-Bromophenyl)ethylamine is utilized as an important raw material in organic synthesis. It also serves as an intermediate in organic synthesis and pharmaceuticals and agrochemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(S)-(-)-1-(4-Bromophenyl)ethylamine is utilized as an important raw material in organic synthesis. It also serves as an intermediate in organic synthesis and pharmaceuticals and agrochemicals.

Notes
Air sensitive. Keep the container tightly closed in a dry and well-ventilated place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 27298-97-1
Molecular Formula C8H11BrN
Molecular Weight (g/mol) 201.09
MDL Number MFCD00066026
InChI Key SOZMSEPDYJGBEK-LURJTMIESA-O
Synonym s---1-4-bromophenyl ethylamine, s-1-4-bromophenyl ethanamine, s---4-bromo-alpha-phenylethylamine, s-1-4-bromophenyl ethylamine, 1s-1-4-bromophenyl ethanamine, s-1-4-bromophenyl-ethylamine, 1s-1-4-bromophenyl ethan-1-amine, s---1-4-bromo-phenyl-ethylamine, benzenemethanamine,4-bromo-a-methyl-, as, s-1-4-bromophenyl ethanamine-hcl
PubChem CID 852999
SMILES C[C@H]([NH3+])C1=CC=C(Br)C=C1

Specifications

Melting Point -25°C (neat)
Density 1.371
Boiling Point 140°C to 145°C (30 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.567
Quantity 5 g
UN Number UN2735
Beilstein 5729988
Sensitivity Air Sensitive
IUPAC Name (1S)-1-(4-bromophenyl)ethan-1-aminium
Formula Weight 200.08
Percent Purity 99%
Assay ee 98%
Chemical Name or Material (S)-(-)-1-(4-Bromophenyl)ethylamine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.