missing translation for 'onlineSavingsMsg'
Learn More

(S)-(-)-1-(4-Bromophenyl)ethyl isocyanate, 97%, Thermo Scientific™

Product Code. 15459159
Click to view available options
Quantity:
1 g
Unit Size:
1g
This item is not returnable. View return policy

Product Code. 15459159

Brand: Thermo Scientific Alfa Aesar L20322.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 149552-52-3
Melting Point 30°C to 32°C
Boiling Point 145°C to 147°C (7 mmHg)
Molecular Formula C9H8BrNO
MDL Number MFCD05664122
Quantity 1 g
UN Number UN2811
Sensitivity Moisture sensitive
Synonym s---1-4-bromophenyl ethyl isocyanate, 1-bromo-4-1s-1-isocyanatoethyl benzene, s-1-4-bromophenyl ethyl isocyanate, s-1-bromo-4-1-isocyanatoethyl benzene, s---1-4-bromophenyl ethylisocyanate, 1-bromo-4-s-1-isocyanato-ethyl-benzene, benzene,1-bromo-4-1s-1-isocyanatoethyl
InChI Key AKVAGWOVWQDTAF-ZETCQYMHSA-N
SMILES CC(C1=CC=C(C=C1)Br)N=C=O
IUPAC Name 1-bromo-4-[(1S)-1-isocyanatoethyl]benzene
Molecular Weight (g/mol) 226.073
PubChem CID 7568842
Formula Weight 226.08
Percent Purity 97%
Chemical Name or Material (S)-(-)-1-(4-Bromophenyl)ethyl isocyanate
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.