missing translation for 'onlineSavingsMsg'
Learn More

(S)-(-)-1-(3-Methoxyphenyl)ethylamine, ChiPros 99+%, ee 99+%

Product Code. 11353007
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11353007 1 g 1g
11363007 5 g 5g
11373007 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11353007 Supplier Thermo Scientific Alfa Aesar Supplier No. L16324.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 82796-69-8 | C9H13NO | 151.209 g/mol

Reatant in the synthesis of the (R)-atropisomer, rivastigmine tartrate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Reatant in the synthesis of the (R)-atropisomer, rivastigmine tartrate.

Solubility
Soluble in water (10g/L).

Notes
Air sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, air.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 82796-69-8
Molecular Formula C9H13NO
Molecular Weight (g/mol) 151.209
MDL Number MFCD00671656
InChI Key CJWGCBRQAHCVHW-ZETCQYMHSA-N
Synonym s-1-3-methoxyphenyl ethanamine, s-1-3-methoxyphenyl ethylamine, 1s-1-3-methoxyphenyl ethanamine, s-3-1-aminoethyl anisole, 1s-1-3-methoxyphenyl ethan-1-amine, s---1-3-methoxyphenyl ethylamine, r-+-1-3-methoxyphenyl ethylamine, chipros, ee, ksc496s2j, s-3-methoxyphenyl ethylamine, 1s-1-3-methoxyphenyl ethylamine
PubChem CID 7020761
IUPAC Name (1S)-1-(3-methoxyphenyl)ethanamine
SMILES CC(C1=CC(=CC=C1)OC)N

Specifications

Melting Point 5°C
Density 1.023
Boiling Point 66°C (0.38 mmHg)
Flash Point >100°C (212°F)
Refractive Index 1.5335
Quantity 1 g
UN Number UN2735
Sensitivity Air Sensitive
Solubility Information Soluble in water (10g/L).
Optical Rotation −27° (Neat)
Formula Weight 151.21
Percent Purity ≥99%
Assay ee ≥99%
Chemical Name or Material (S)-(-)-1-(3-Methoxyphenyl)ethylamine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.