missing translation for 'onlineSavingsMsg'
Learn More

(S)-(+)-1,3-Butanediol, 98%

Product Code. 10147353 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10147353 25 g Glass bottle 25g
10440491 1 g Glass bottle 1g
10174650 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10147353 Supplier Thermo Scientific Acros Supplier No. 217970250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 24621-61-2 | C4H10O2 | 90.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 24621-61-2
Molecular Formula C4H10O2
Molecular Weight (g/mol) 90.12
MDL Number MFCD00064278
InChI Key PUPZLCDOIYMWBV-BYPYZUCNSA-N
Synonym s-+-1,3-butanediol, s-butane-1,3-diol, 3s-butane-1,3-diol, s-1,3-butanediol, s-+-butane-1,3-diol, 1,3-butanediol, 3s, bu2, 1,3-butanediol, s, pubchem6722
PubChem CID 446973
ChEBI CHEBI:52688
IUPAC Name (3S)-butane-1,3-diol
SMILES CC(CCO)O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Colorless to Yellow
Density 1.0000g/mL
Boiling Point 109.0°C (14.0 mmHg)
Flash Point 121°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3CH(OH)CH2CH2OH
Refractive Index 1.4385 to 1.4405
Quantity 25 g
Specific Gravity 1
Specific Rotation Condition +30° (20°C c=1,C2H5OH)
Specific Rotation + 30.00
Solubility Information Solubility in water: soluble. Other solubilities: soluble in acetone, methyl ethyl ketone, ethanol, slightly soluble in ether, practically insoluble in aliphatic hydrocarbons,, benzene, toluene and ccl4
Formula Weight 90.12
Percent Purity 98%
Physical Form Viscous Liquid
Chemical Name or Material (S)-(+)-1, 3-Butanediol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.