missing translation for 'onlineSavingsMsg'
Learn More

(S)-1-(3-Bromophenyl)ethylamine, ChiPros™ 99%, ee 98+%

Product Code. 11471640
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11471640 5 g 5g
11461640 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11471640 Supplier Thermo Scientific Alfa Aesar Supplier No. H26998.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 139305-96-7 | C8H10BrN | 200.079 g/mol

(S)-1-(3-Bromophenyl)ethylamine is involved in the Ullmann coupling reaction with N-H containing heteroarenes using copper iodide as a catalyst.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(S)-1-(3-Bromophenyl)ethylamine is involved in the Ullmann coupling reaction with N-H containing heteroarenes using copper iodide as a catalyst.

Solubility
Immiscible with water.

Notes
Incompatible with strong oxidizing agents, carbon monoxide and strong acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 139305-96-7
Molecular Formula C8H10BrN
Molecular Weight (g/mol) 200.079
MDL Number MFCD00671638
InChI Key LIBZHYLTOAGURM-LURJTMIESA-N
Synonym s-1-3-bromophenyl ethanamine, s-1-3-bromophenyl ethylamine, 1s-1-3-bromophenyl ethanamine, 1s-1-3-bromophenyl ethan-1-amine, s-3-bromo-alpha-methylbenzylamine, 1s-1-3-bromophenyl ethylamine, s---3-bromo alpha-methylbenzylamine, s-m-bromo-, a-methylbenzylamine, s-3-bromo-
PubChem CID 9952938
IUPAC Name (1S)-1-(3-bromophenyl)ethanamine
SMILES CC(C1=CC(=CC=C1)Br)N

Specifications

Density 1.33
Boiling Point 96°C (4 mmHg)
Flash Point >110°C (230°F)
Quantity 5 g
UN Number UN2735
Sensitivity Air Sensitive
Solubility Information Immiscible with water.
Optical Rotation −22° (Neat)
Formula Weight 200.08
Percent Purity 99%
Assay ee ≥98%
Chemical Name or Material (S)-1-(3-Bromophenyl)ethylamine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.