missing translation for 'onlineSavingsMsg'
Learn More

(S)-(+)-1,2-Propanediol, 97%, ACROS Organics™

Product Code. 10029001 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
:
5g; Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10029001 5g; Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10029001 Supplier Thermo Scientific Acros Supplier No. 365300050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 4254-15-3
CAS Min % 96.0
CAS Max % 100.0
Melting Point -59.0°C
Color Colorless
Density 102g/mL
Boiling Point 186.0°C to 188.0°C (765.0 mmHg)
Flash Point 102°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Molecular Formula C3H8O2
Linear Formula CH3CH(OH)CH2OH
Refractive Index 1.4310 to 1.4350
MDL Number MFCD00004539
Quantity 5 g
Merck Index 15, 7968
Specific Rotation Condition + 16.00 (20.00°C c=neat)
Specific Rotation + 16.00
Synonym s-+-1,2-propanediol, s-propane-1,2-diol, s-1,2-propanediol, 2s-propane-1,2-diol, s-propylene glycol, s-+-propylene glycol, s-+-1,2-dihydroxypropane, propylene glycol #, pgo
Solubility Information Solubility in water: soluble in water.
InChI Key DNIAPMSPPWPWGF-UHFFFAOYNA-N
SMILES CC(O)CO
IUPAC Name propane-1,2-diol
Molecular Weight (g/mol) 76.10
PubChem CID 439846
ChEBI CHEBI:29002
Formula Weight 76.1
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material (S)-(+)-1, 2-Propanediol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.