missing translation for 'onlineSavingsMsg'
Learn More

(S)-1,2,3,4-Tetrahydroisoquinoline-3-acetic acid hydrochloride, 95%, Thermo Scientific™

Product Code. 15449595
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15449595 250 mg 250mg
15439595 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15449595 Supplier Thermo Scientific Alfa Aesar Supplier No. H52111.MD

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
It is used as an active pharmaceutical intermediate. It is also an important organic intermediate used in agrochemicals, pharmaceuticals and dyestuff fields.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 270082-22-9
Molecular Formula C11H14ClNO2
Molecular Weight (g/mol) 227.69
MDL Number MFCD01861030
InChI Key SWYPWAIQEURSFY-PPHPATTJSA-N
Synonym s-2-tetrahydroisoquinoline acetic acid hydrochloride, s-2-1,2,3,4-tetrahydroisoquinolin-3-yl acetic acid hydrochloride, s-1,2,3,4-tetrahydro-3-isoquinolineacetic acid hydrochloride, 2-3s-1,2,3,4-tetrahydroisoquinolin-3-yl acetic acid hydrochloride, s-2-tetrahydroisoquinoline acetic acid-hcl, 3s-1,2,3,4-tetrahydroisoquinolin-3-ylacetic acid hydrochloride, h-tqa-ohcl, h-tqa-oh.hcl, s-1,2,3,4-tetrahydroisoquinoline-3-acetic acid hydrochloride
PubChem CID 2761613
IUPAC Name 2-[(3S)-1,2,3,4-tetrahydroisoquinolin-3-yl]acetic acid;hydrochloride
SMILES [H+].[Cl-].OC(=O)C[C@@H]1CC2=CC=CC=C2CN1

Specifications

Quantity 250 mg
Formula Weight 227.69
Percent Purity 95%
Chemical Name or Material (S)-1,2,3,4-Tetrahydroisoquinoline-3-acetic acid hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.