missing translation for 'onlineSavingsMsg'
Learn More

(S)-1,2,3,4-Tetrahydro-1-naphthoic Acid, TRC

Product Code. 30410407
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30410407 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30410407 Supplier Toronto Research Chemicals Supplier No. TRCT294115250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (S)-1,2,3,4-Tetrahydro-1-naphthoic Acid
CAS 85977-52-2
Molecular Formula C11H12O2
Quantity 250 mg
Synonym (1S)-1,2,3,4-Tetrahydro-1-naphthalenecarboxylic Acid, D-(-)-1,2,3,4-Tetrahydro-1-naphthoic Acid
InChI Formula InChI=1S/C11H12O2/c12-11(13)10-7-3-5-8-4-1-2-6-9(8)10/h1-2,4,6,10H,3,5,7H2,(H,12,13)/t10-/m0/s1
SMILES OC(=O)[C@H]1CCCc2ccccc12
IUPAC Name (1S)-1,2,3,4-tetrahydronaphthalene-1-carboxylic acid
Molecular Weight (g/mol) 176.21
Formula Weight 176.0837
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.