missing translation for 'onlineSavingsMsg'
Learn More

(S)-1,2,3,4-Tetrahydro-1-naphthoic acid, 98%

Product Code. 11486540
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11486540 1 g 1g
15403737 250 mg 250mg
11496540 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11486540 Supplier Thermo Scientific Alfa Aesar Supplier No. H27612.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 85977-52-2 | C11H12O2 | 176.215 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 85977-52-2
Molecular Formula C11H12O2
Molecular Weight (g/mol) 176.215
MDL Number MFCD07368365
InChI Key VDLWTJCSPSUGOA-JTQLQIEISA-N
Synonym s-1,2,3,4-tetrahydro-1-naphthoic acid, s-1,2,3,4-tetrahedro-naphthoic acid, 1s-1,2,3,4-tetrahydronaphthalene-1-carboxylic acid, s-1,2,3,4-tetrahydro-naphthoic acid, 1s-tetralin-1-carboxylic acid, s---1,2,3,4-tetrahydronaphthoic acid, s-1,2,3,4-tetrahydronaphthalene-1-carboxylic acid, pubchem11869, s-1, 2, 3, 4-tetrahydronaphthoic acid, 1s-1,2,3,4-tetrahydro-1-naphthoic acid
PubChem CID 24820460
IUPAC Name (1S)-1,2,3,4-tetrahydronaphthalene-1-carboxylic acid
SMILES C1CC(C2=CC=CC=C2C1)C(=O)O

Specifications

Melting Point 55°C to 57°C
Quantity 1 g
Optical Rotation −12° (c=1 in Ethanol)
Formula Weight 176.22
Percent Purity 98%
Chemical Name or Material (S)-1,2,3,4-Tetrahydro-1-naphthoic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.