missing translation for 'onlineSavingsMsg'
Learn More

(S)-(-)-1,1'-Bi-2-naphthol, 99%

Product Code. 10444335 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10444335 25 g 25g
10591471 1 g 1g
10441173 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10444335 Supplier Thermo Scientific Acros Supplier No. 224310250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 18531-99-2 | C20H14O2 | 286.32 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18531-99-2
InChI Key PPTXVXKCQZKFBN-UHFFFAOYSA-N
Synonym 1,1'-bi-2-naphthol, s---1,1'-bi-2-naphthol, r-+-1,1'-bi-2-naphthol, 1,1'-binaphthalene-2,2'-diol, binol, 1,1'-binaphthyl-2,2'-diol, 2,2'-dinaphthol, beta-binaphthol, r-1,1'-bi-2-naphthol, s-1,1'-binaphthalene-2,2'-diol
PubChem CID 11762
IUPAC Name 1-(2-hydroxynaphthalen-1-yl)naphthalen-2-ol
SMILES C1=CC=C2C(=C1)C=CC(=C2C3=C(C=CC4=CC=CC=C43)O)O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 205°C to 211°C
Color White to Gray
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Glass bottle
Molecular Formula C20H14O2
Linear Formula HOC10H6C10H6OH
Quantity 25 g
Fieser 16,132
Merck Index 15, 1230
Specific Rotation Condition −35.50° (20°C c=1,THF)
Specific Rotation -35.5
Solubility Information Solubility in water: insoluble
Molecular Weight (g/mol) 286.32
Formula Weight 286.32
Percent Purity 99%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material (S)-(-)-1, 1'-Bi-2-naphthol, 99%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.