missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ S 07662
Description
S 07662 is an inverse agonist of the constitutive androstane receptor (CAR). Inhibits CAR activity via recruitment of nuclear receptor co-repressor (NCoR). Suppresses CITCO -induced CYP2B6 gene expression in human primary hepatocytes.
Specifications
Specifications
| Purity | >98% |
| Quantity | 50 mg |
| Product Type | S 07662 |
| Molecular Weight (g/mol) | 300.38 |
| Inhibitors | S 07662 |
For Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?