missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RyR1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5115857
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total RYR1.
Ryanodine receptors have been shown to play critical roles in the intracellular Ca2+ signaling occurring during cell activation in muscle cells and non-muscled cells. Ryanodine receptor, ~500 kDa, can be isolated from a variety of cell types such as lymphocytes, macrophages, granulocytes, fibroblasts, epithelial, endothelial cells, skeletal muscles, cardiac muscle and brain tissues.
Specifications
| RyR1 | |
| Polyclonal | |
| Unconjugated | |
| RYR1 | |
| AI528790; calcium release channel; calcium release channel (ryanodine receptor 1); calcium release channel protein; CCO; central core disease of muscle; CRC; CRC1, RYR1; halothane; LOW QUALITY PROTEIN: ryanodine receptor 1; MHS; MHS1; porcine stress syndrome; PPP1R137; protein phosphatase 1, regulatory subunit 137; ryanodine receptor; ryanodine receptor 1; ryanodine receptor 1 (skeletal); ryanodine receptor 1, skeletal muscle; ryanodine receptor 1-like, skeletal muscle; RYDR; Ryr; Ryr1; RYR-1; Ryr1l; sarcoplasmic reticulum calcium release channel; skeletal muscle calcium release channel; Skeletal muscle ryanodine receptor; skeletal muscle-type ryanodine receptor; skrr; type 1 ryanodine receptor; type 1-like ryanodine receptor; unnamed protein product | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 6261 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide | |
| P21817 | |
| RYR1 | |
| A synthesized peptide derived from human RYR1(Accession P21817), corresponding to amino acid residues E1113-L1163. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction