missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RYR1 Mouse anti-Human, Mouse, Clone: 1B4E12, Proteintech
Mouse Monoclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | RYR1 |
|---|---|
| Clone | 1B4E12 |
| Concentration | 1 mg/mL |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16888505
|
Proteintech
66539-1-IG-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16878505
|
Proteintech
66539-1-IG-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Ryanodine receptors have been shown to play critical roles in the intracellular Ca2+ signaling occurring during cell activation in muscle cells and non-muscled cells. Ryanodine receptor, ∽500 kDa, can be isolated from a variety of cell types such as lymphocytes, macrophages, granulocytes, fibroblasts, epithelial, endothelial cells, skeletal muscles, cardiac muscle and brain tissues.Specifications
| RYR1 | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| CCO, MHS, MHS1, Ryanodine receptor 1, RYDR, RYR, RYR 1, RYR1, SKRR | |
| RYR1 | |
| IgG1 | |
| Protein G | |
| Antibody |
| 1B4E12 | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Mouse, Human | |
| E9PZQ0, P21817 | |
| 20190, 6261 | |
| RYR1 Fusion Protein Ag25120 | |
| Primary | |
| -20°C | |
| RYR1 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title