missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RYK Recombinant Rabbit Monoclonal Antibody (24GB5080)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA557212
This item is not returnable.
View return policy
Description
RYK Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
RYK is an atypical member of the family of growth factor receptor protein tyrosine kinases, differing from other members at a number of conserved residues in the activation and nucleotide binding domains. This gene product belongs to a subfamily whose members do not appear to be regulated by phosphorylation in the activation segment. It has been suggested that mediation of biological activity by recruitment of a signaling-competent auxiliary protein may occur through an as yet uncharacterized mechanism. A nine nucleotide insertion in some transcripts results in the SLG variant. It is not established whether this is a product of alternative splicing or a second gene, since evidence for a second gene or pseudogene on chromosome 17 exists.
Specifications
| RYK | |
| Recombinant Monoclonal | |
| Unconjugated | |
| RYK | |
| AW536699; D3S3195; ERK3; ERK-3; growth factor receptor; hydroxyaryl-protein kinase; JTK5; JTK5A; JTK5A protein tyrosine kinase; kinase VIK; Met-related kinase; Mrk; NYK-R; receptor-like tyrosine kinase; RYK; RYK1; tyrosine-protein kinase RYK; Vik | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 6259 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 24GB5080 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P34925 | |
| RYK | |
| A synthesized peptide derived from human RYK (aa 575-589). | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction