missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RYK Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA130319
This item is not returnable.
View return policy
Description
Detects a band of approximately 67.5 kDa (predicted molecular weight: 72 kDa). Positive Control: Jurkat cell lysate. Store as concentrated solution. Centrifuge briefly prior to opening vial.
RYK is an atypical member of the family of growth factor receptor protein tyrosine kinases, differing from other members at a number of conserved residues in the activation and nucleotide binding domains. This gene product belongs to a subfamily whose members do not appear to be regulated by phosphorylation in the activation segment. It has been suggested that mediation of biological activity by recruitment of a signaling-competent auxiliary protein may occur through an as yet uncharacterized mechanism. A nine nucleotide insertion in some transcripts results in the SLG variant. It is not established whether this is a product of alternative splicing or a second gene, since evidence for a second gene or pseudogene on chromosome 17 exists.
Specifications
| RYK | |
| Polyclonal | |
| Unconjugated | |
| RYK | |
| AW536699; D3S3195; ERK3; ERK-3; growth factor receptor; hydroxyaryl-protein kinase; JTK5; JTK5A; JTK5A protein tyrosine kinase; kinase VIK; Met-related kinase; Mrk; NYK-R; receptor-like tyrosine kinase; RYK; RYK1; tyrosine-protein kinase RYK; Vik | |
| Rabbit | |
| Protein A | |
| RUO | |
| 140585, 20187, 6259 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.40 mg/mL | |
| PBS with 0.09% sodium azide | |
| P34925, Q01887 | |
| RYK | |
| KLH conjugated synthetic peptide between 160-190 amino acids from human RYK. | |
| 400 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction