missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ryk Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-37915
This item is not returnable.
View return policy
Description
Ryk Polyclonal specifically detects Ryk in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Ryk | |
| Polyclonal | |
| Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
| P34925 | |
| RYK | |
| This antibody was developed against a recombinant protein corresponding to amino acids: SMKRIELDDSISASSSSQGLSQPSTQTTQYLRADTPNNATPITSSLGYPTLRIEKNDLRSVTLLEAKGKVKDIAISRERITLKDVLQEGTFGRIFHGILIDEKDPNKEKQAFVKTVKDQASEIQVTMMLTESCKLRGLHHRNLLPITH | |
| 0.1 mL | |
| Protein Kinase | |
| 6259 | |
| Human | |
| IgG |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| D3S3195, EC 2.7.10, EC 2.7.10.1, hydroxyaryl-protein kinase, JTK5, JTK5A, JTK5A protein tyrosine kinase, RYK receptor-like tyrosine kinase, RYK1, tyrosine-protein kinase RYK | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction