missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RYBP/DEDAF Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-35449-100ul
This item is not returnable.
View return policy
Description
RYBP/DEDAF Polyclonal antibody specifically detects RYBP/DEDAF in Mouse,Rat samples. It is validated for ELISA,Western Blot
Specifications
| RYBP/DEDAF | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, ELISA | |
| AAP1, APAP-1, Apoptin-associating protein 1, Death effector domain-associated factor, DEDAFYY1 and E4TF1-associated factor 1, RING1 and YY1 binding protein, RING1 and YY1-binding protein, ring1 interactor RYBP, YEAF1DED-associated factor, YY1 and E4TF1 associated factor 1 | |
| A synthetic peptide corresponding to a sequence within amino acids 100 to the C-terminus of human RYBP/DEDAF (NP_036366.3).,, Sequence:, PSVTKKNTNKKTKPKSDILKDPPSEANSIQSANATTKTSETNHTSRPRLKNVDRSTAQQLAVTVGNVTVIITDFKEKTRSSSTSSSTVTSSAGSEQQNQSSSGSESTDKGSSRSSTPKGDMSAVNDESF | |
| 100 μL | |
| Apoptosis | |
| 23429 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction