missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ryanodine Receptor Antibody (34C), PerCP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB300-543PCP
This item is not returnable.
View return policy
Description
Ryanodine Receptor Monoclonal antibody specifically detects Ryanodine Receptor in Human, Mouse, Rat, Amphibian, Bovine, Canine, Fish, Ferret, Mink, Primate, Rabbit, Sheep samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Neutralization
Specifications
| Ryanodine Receptor | |
| Monoclonal | |
| PerCP | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat, Amphibian, Bovine | |
| Purified |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Blocking Assay | |
| 34C | |
| Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Block/Neutralize | |
| ARVC2, ARVD2, cardiac muscle ryanodine receptor-calcium release channel, cardiac-type ryanodine receptor, CCO, central core disease of muscle, islet-type ryanodine receptor, kidney-type ryanodine receptor, MHS, MHS1, PPP1R137, protein phosphatase 1, regulatory subunit 137, ryanodine receptor 1, ryanodine receptor 1 (skeletal), ryanodine receptor 2, ryanodine receptor 2 (cardiac), RYDR, RYR, RYR-1, RYR-2, sarcoplasmic reticulum calcium release channel, Skeletal muscle calcium release channel, skeletal muscle ryanodine receptor, SKRR, type 1-like ryanodine receptor, type 2 ryanodine receptor, VTSIP | |
| Partially purified chicken pectoral muscle ryanodine receptor. | |
| 0.1 mL | |
| Neuroscience | |
| 6261 | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction