missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ryanodine Receptor Antibody (34C), DyLight 680, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB300-543FR
This item is not returnable.
View return policy
Description
Ryanodine Receptor Monoclonal specifically detects Ryanodine Receptor in Human, Mouse, Rat, Amphibian, Bovine, Canine, Fish, Ferret, Mink, Primate, Rabbit, Sheep samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifications
| Ryanodine Receptor | |
| Monoclonal | |
| DyLight 680 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| RYR1 | |
| Partially purified chicken pectoral muscle ryanodine receptor. | |
| 0.1 mL | |
| Neuroscience | |
| This antibody detects ryanodine receptor Ryanodine Receptor-1 and Ryanodine Receptor-2 isoforms in amphibian, chicken, bovine, canine, fish, human, non-human primate, mouse, rabbit, mink, rat and sheep tissues. In chicken, it detects the alpha, beta and cardiac isoforms. This antibody also detects Ryanodine Receptor-3 in mouse cells. In frog, it detects the alpha and beta isoforms and in fish it detects the alpha isoform. | |
| Store at 4°C in the dark. |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
| 34C | |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen | |
| ARVC2, ARVD2, cardiac muscle ryanodine receptor-calcium release channel, cardiac-type ryanodine receptor, CCO, central core disease of muscle, islet-type ryanodine receptor, kidney-type ryanodine receptor, MHS, MHS1, PPP1R137, protein phosphatase 1, regulatory subunit 137, ryanodine receptor 1, ryanodine receptor 1 (skeletal), ryanodine receptor 2, ryanodine receptor 2 (cardiac), RYDR, RYR, RYR-1, RYR-2, sarcoplasmic reticulum calcium release channel, Skeletal muscle calcium release channel, skeletal muscle ryanodine receptor, SKRR, type 1-like ryanodine receptor, type 2 ryanodine receptor, VTSIP | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat, Amphibian, Bovine, Canine, Fish, Ferret, Mink, Primate, Rabbit, Sheep | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction